Iron(III) oxide 98
Supplier: BeanTown Chemical
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
127425-1KG
127425-5KG
BT127425-1KGEA
57.26
USD
BT127425-1KG
BT127425-5KG
Iron(III) oxide 98
Iron(III) oxide
CAS: 1309-37-1; EC No: 215-168-2; MDL No: MFCD00011008; RTECS: NO7400000
-325 Mesh Powder; Molecular Formula: Fe2O3 ; MW: 159.69
- RTECS: NO7400000
- -325 Mesh Powder
Formula:
Fe₂O₃ MW: 159.69 g/mol Melting Pt: 1538 |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 13,04058 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...