2-(Benzyloxy)ethanol ≥98%
Supplier: BeanTown Chemical
CAS: 622-08-2; EC No: 210-719-3; MDL No: MFCD00002868; RTECS: KJ7550000
Liquid; Linear Formula: C6H5CH2OCH2CH2OH; Molecular Formula: C9H12O2; MW: 152.19
Boiling Point: 265°; Flash point: 110°C (230°F)
Density (g/mL): 1.071; Refractive Index: 1.521
- RTECS: KJ7550000
- Refractive index: 1.521
- Liquid
Formula:
C₉H₁₂O₂ Boiling Pt: 265 °C Density: 1.071 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00002868 CAS Number: 622-08-2 EINECS: 210-719-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...