2-Bromo-4-methylpyridine ≥96%

Supplier: BeanTown Chemical

Synonyms: 2-Bromo-4-picoline

127395-1G 127395-25G 127395-5G
BT127395-1GEA 38 USD
BT127395-1G BT127395-25G BT127395-5G
2-Bromo-4-methylpyridine ≥96%
2-Bromo-4-methylpyridine

CAS: 4926-28-7; MDL No: MFCD00082590 Liquid; Molecular Formula: C6H6BrN ; MW: 172.02 Boiling Point: 87°/10 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.545; Refractive Index: 1.561


  • Refractive index: 1.561
  • Liquid

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 87 °C/10 mmHg
Density: 1.545 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00082590
CAS Number: 4926-28-7

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR