2-Hydroxynicotinic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

127365-100G 127365-25G 127365-500G
BT127365-100GEA 159 USD
BT127365-100G BT127365-25G BT127365-500G
2-Hydroxynicotinic acid ≥99%
2-Hydroxynicotinic acid

CAS: 609-71-2; EC No: 210-198-2; MDL No: MFCD00010100 Powder; Molecular Formula: C6H4NO3; MW: 139.11 Melting Point: 258-261°


  • Powder

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 258…261 °C
MDL Number: MFCD00010100
CAS Number: 609-71-2
EINECS: 210-198-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR