3-Methylindole ≥99%

Supplier: BeanTown Chemical

Synonyms: Skatol, 3-Methyl-1H-indole, β-Methylindole, Skatole

127355-25G 127355-5G
BT127355-25GEA 83.98 USD
BT127355-25G BT127355-5G
3-Methylindole ≥99%
3-Methylindole

CAS: 83-34-1; EC No: 201-471-7; MDL No: MFCD00005627; RTECS: NM0350000 Flakes; Molecular Formula: C9H9N; MW: 131.17 Melting Point: 92-97°; Boiling Point: 265-267°; Flash point: 132°C (270°F) Light Sensitive


  • RTECS: NM0350000
  • Light Sensitive
  • Flakes

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 265…267 °C
Melting Pt: 92…97 °C
Flash Pt: 132 °C (270 °F)
MDL Number: MFCD00005627
CAS Number: 83-34-1
EINECS: 201-471-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR