Boc-Dap(Cbz)-OH

Supplier: BeanTown Chemical

Synonyms: Boc-Dap(Z)-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-3-(benzyloxycarbonylamino)propionic acid

127340-1G 127340-250MG
BT127340-1GEA 627 USD
BT127340-1G BT127340-250MG
Boc-Dap(Cbz)-OH
Boc-Dap(Cbz)-OH

CAS: 65710-57-8; MDL No: MFCD08275856 Solid; Molecular Formula: C16H22N2O6; MW: 338.36


  • Solid

Formula: C₁₆H₂₂N₂O₆
MW: 338.36 g/mol
MDL Number: MFCD08275856
CAS Number: 65710-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR