N-Methylphthalimide ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Methylisoindoline-1,3-dione

127330-25G 127330-5G
BT127330-25GEA 71 USD
BT127330-25G BT127330-5G
N-Methylphthalimide ≥98%
N-Methylphthalimide

CAS: 550-44-7; EC No: 208-982-4; MDL No: MFCD00023063; RTECS: TI5602700 Powder; Molecular Formula: C9H7NO2 ; MW: 161.16 Melting Point: 129-132°


  • RTECS: TI5602700
  • Powder

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD00023063
CAS Number: 550-44-7
EINECS: 208-982-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR