O-Benzyl-L-serine ≥99%

Supplier: BeanTown Chemical

Synonyms: H-Ser(Bzl)-OH

127275-1G 127275-25G 127275-5G
BT127275-1GEA 43 USD
BT127275-1G BT127275-25G BT127275-5G
O-Benzyl-L-serine ≥99%
O-Benzyl-L-serine

CAS: 4726-96-9; EC No: 225-220-6; MDL No: MFCD00065937 Solid; Molecular Formula: C10H13NO3; MW: 195.22 Melting Point: 227° (decomposes)


  • Solid

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 227 °C (dec.)
MDL Number: MFCD00065937
CAS Number: 4726-96-9
EINECS: 225-220-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR