Tin(IV) acetate
Supplier: BeanTown Chemical
127250-25G
127250-5G
BT127250-25GEA
600
USD
BT127250-25G
BT127250-5G
Tin(IV) acetate
Tin(IV) acetate
CAS: 2800-96-6; MDL No: MFCD00014976
Crystalline; Linear Formula: Sn(OOCCH3)4; MW: 354.89
Melting Point: 232-233°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₈H₁₂O₈Sn MW: 354.89 Melting Pt: 232…233 °C |
MDL Number:
MFCD00014976 CAS Number: 2800-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...