Methyl-2-chloroisonicotinate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate
127235-1G
127235-5G
BT127235-1GEA
40
USD
BT127235-1G
BT127235-5G
Methyl-2-chloroisonicotinate ≥97%
Methyl 2-chloroisonicotinate
CAS: 58481-11-1; MDL No: MFCD01765409
Solid; Molecular Formula: C7H6ClNO2; MW: 171.58
Melting Point: 32-36°; Flash point:
- Solid
Formula:
C₇H₆ClNO₂ MW: 171.5810 g/mol Melting Pt: 32…36 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD01765409 CAS Number: 58481-11-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...