Diethyl difluoromalonate ≥97%
Supplier: BeanTown Chemical
127225-1G
127225-5G
BT127225-1GEA
74
USD
BT127225-1G
BT127225-5G
Diethyl difluoromalonate ≥97%
Diethyl difluoromalonate
CAS: 680-65-9; MDL No: MFCD00221617
Liquid; Molecular Formula: C7H10F2O4; MW: 196.15
Boiling Point: 94-95°/23 mmHg
Density (g/mL): 1.179
- Liquid
Formula:
C₇H₁₀F₂O₄ MW: 196.15 g/mol Boiling Pt: 94…95 °C/23 mmHg Density: 1.179 g/mL |
MDL Number:
MFCD00221617 CAS Number: 680-65-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...