Boc-L-pyroglutamic acid methyl ester ≥95%
Supplier: BeanTown Chemical
Synonyms:
Methyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Methyl N-Boc-L-pyroglutamate, Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, Boc-Pyr-OMe, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-Boc-L-pyroglutamic acid methyll ester, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid methyll ester, 1-(tert-Butyl) 2-methyl (S)-5-oxopyrrolidine-1,2-dicarboxylate, Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
CAS: 108963-96-8; MDL No: MFCD06809720
Solid; Molecular Formula: C11H17NO5; MW: 243.26
Melting Point: 69-74°
Optical Rotation: [α]22/D -32.0°, c = 0.5 in chloroform
- Optical rotation: [α]22/D -32.0°, c = 0.5 in chloroform
- Solid
Formula:
C₁₁H₁₇NO₅ MW: 243.26 g/mol Melting Pt: 69…74 °C |
MDL Number:
MFCD06809720 CAS Number: 108963-96-8 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...