Polyvinyl alcohol 87-89%, high molecular weight, hydrolysed

Supplier: BeanTown Chemical

127165-100G 127165-25G 127165-500G
BT127165-100GEA 39.17 USD
BT127165-100G BT127165-25G BT127165-500G
Polyvinyl alcohol 87-89%, high molecular weight, hydrolysed
Polyvinyl alcohol

CAS: 9002-89-5; MDL No: MFCD00081922 Crystalline; Linear Formula: [-CH2CH(OH)-]n Melting Point: 200° Density (g/mL): 1.269


  • Crystalline

Formula: [-CH₂CH(OH)-]n
Melting Pt: 200 °C
Density: 1.269 g/mL
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR