Ethyl enantate ≥98%

Supplier: BeanTown Chemical

127145-100G 127145-500G
BT127145-100GEA 25 USD
BT127145-100G BT127145-500G
Ethyl enantate ≥98%
Ethyl enantate

CAS: 106-30-9; EC No: 203-382-9; MDL No: MFCD00009538; RTECS: MJ2087000 Liquid; Linear Formula: CH3(CH2)5COOCH2CH3; Molecular Formula: C9H18O2; MW: 158.24 Melting Point: -66°; Boiling Point: 188-189°; Flash point: 74°C (165°F) Density (g/mL): 0.87; Refractive Index: 1.412


  • RTECS: MJ2087000
  • Refractive index: 1.412
  • Liquid

Formula: C₉H₁₈O₂
Boiling Pt: 188…189 °C
Melting Pt: –66 °C
Density: 0.87 g/mL
Flash Pt: 74 °C (165 °F)
MDL Number: MFCD00009538
CAS Number: 106-30-9
EINECS: 203-382-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR