4-Quinolinecarbaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde

127135-25G 127135-5G
BT127135-25GEA 348 USD
BT127135-25G BT127135-5G
4-Quinolinecarbaldehyde ≥97%
4-Quinolinecarbaldehyde

CAS: 4363-93-3; EC No: 224-453-0; MDL No: MFCD00006781 Solid; Molecular Formula: C10H7NO; MW: 157.17 Melting Point: 48-52°; Flash point: 110°C (230°F) Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 48…52 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR