2-Bromonaphthalene ≥98%

Supplier: BeanTown Chemical

127125-100G 127125-25G 127125-5G
BT127125-100GEA 548 USD
BT127125-100G BT127125-25G BT127125-5G
2-Bromonaphthalene ≥98%
2-Bromonaphthalene

CAS: 580-13-2; EC No: 209-452-5; MDL No: MFCD00004051 Powder; Molecular Formula: C10H7Br; MW: 207.07 Melting Point: 52-55°; Boiling Point: 281-282°; Flash point: 110°C (230°F)


  • Powder

Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 281…282 °C
Melting Pt: 52…55 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004051
CAS Number: 580-13-2
EINECS: 209-452-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR