2,6-Dichloro-4-iodopyridine ≥97%
Supplier: BeanTown Chemical
127090-1G
127090-5G
BT127090-1GEA
81
USD
BT127090-1G
BT127090-5G
2,6-Dichloro-4-iodopyridine ≥97%
2,6-Dichloro-4-iodopyridine
CAS: 98027-84-0; MDL No: MFCD07368400
Solid; Molecular Formula: C5H2Cl2IN; MW: 273.89
Melting Point: 161-165°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₅H₂CI₂IN MW: 273.89 g/mol Melting Pt: 161…165 °C |
MDL Number:
MFCD07368400 CAS Number: 98027-84-0 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...