Potassium cyanate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Cyanic acid potassium salt
127065-100G
127065-2KG
127065-500G
BT127065-100GEA
29
USD
BT127065-100G
BT127065-2KG
BT127065-500G
Potassium cyanate ≥97%
Potassium cyanate
CAS: 590-28-3; EC No: 209-676-3; MDL No: MFCD00011411
Crystalline; Linear Formula: KOCN; MW: 81.12
Melting Point: 700-900° (decomposes)
Density (g/mL): 2.056
- Crystalline
Formula:
CKNO MW: 81.12 g/mol Melting Pt: 700…900 °C (dec.) Density: 2.056 g/mL |
MDL Number:
MFCD00011411 CAS Number: 590-28-3 EINECS: 209-676-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...