3-Ethyl-4-methyl-3-pyrrolin-2-one ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 1,5-Dihydro-3-ethyl-4-methyl-2H-pyrrol-2-one

127020-1G 127020-250MG 127020-5G
BT127020-1GEA 37 USD
BT127020-1G BT127020-250MG BT127020-5G
3-Ethyl-4-methyl-3-pyrrolin-2-one ≥98%
3-Ethyl-4-methyl-3-pyrrolin-2-one

CAS: 766-36-9; MDL No: MFCD00173861 Powder; Molecular Formula: C7H11NO; MW: 125.17 Melting Point: 100-104°


  • Powder

Formula: C₇H₁₁NO
MW: 125.17 g/mol
Melting Pt: 100…104 °C
MDL Number: MFCD00173861
CAS Number: 766-36-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR