Methyl-3-hydroxypicolinate ≥95%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-Hydroxy-2-pyridinecarboxylate, 3-Hydroxypyridine-2-carboxylic acid methyl ester, 3-Hydroxypicolinic acid methyll ester, Methyl 3-hydroxypicolinate, 3-Hydroxy-2-pyridinecarboxylic acid methyll ester, Methyl 3-Hydroxypicolinate

126995-1G 126995-5G
BT126995-1GEA 183 USD
BT126995-1G BT126995-5G
Methyl-3-hydroxypicolinate ≥95%
Methyl-3-hydroxypicolinate

CAS: 62733-99-7; MDL No: MFCD00661311 Solid; Molecular Formula: C7H7NO3; MW: 153.14


  • Solid

Formula: C₇H₇NO₃
MDL Number: MFCD00661311
CAS Number: 62733-99-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR