3-Bromo-4-hydroxybenzoic acid ≥98%

Supplier: BeanTown Chemical

126990-10G 126990-50G
BT126990-10GEA 53 USD
BT126990-10G BT126990-50G
3-Bromo-4-hydroxybenzoic acid ≥98%
3-Bromo-4-hydroxybenzoic acid

CAS: 14348-41-5; EC No: 238-299-7; MDL No: MFCD00017547 Solid; Molecular Formula: C7H5BrO3; MW: 217.02 Melting Point: 155-160°


  • Solid

Formula: C₇H₅BrO₃
MW: 217.02 g/mol
Melting Pt: 155…160 °C
MDL Number: MFCD00017547
CAS Number: 14348-41-5
EINECS: 238-299-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR