Magnesium diboride ≥99%

Supplier: BeanTown Chemical

Synonyms: Magnesium boride

126985-100G 126985-25G 126985-5G
BT126985-100GEA 307 USD
BT126985-100G BT126985-25G BT126985-5G
Magnesium diboride ≥99%
Magnesium diboride

CAS: 12007-25-9; EC No: 234-501-2; MDL No: MFCD00049752; RTECS: OM3078000 -100 Mesh Powder; Linear Formula: MgB2; MW: 45.93 Melting Point: 800° (decomposes)


  • RTECS: OM3078000
  • -100 Mesh Powder

Formula: B₂Mg
Melting Pt: 800 °C (dec.)
MDL Number: MFCD00049752
CAS Number: 12007-25-9
EINECS: 234-501-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR