Manganese(III) oxide ≥98%

Supplier: BeanTown Chemical

Synonyms: Manganese oxide (Mn2O3), Dimanganese trioxide

126935-2KG 126935-500G
BT126935-2KGEA 498 USD
BT126935-2KG BT126935-500G
Manganese(III) oxide ≥98%
Manganese(III) oxide

CAS: 1317-34-6; EC No: 215-264-4; MDL No: MFCD00016217 -325 Mesh Powder; Molecular Formula: Mn2O3; MW: 157.86 Melting Point: 1080° (decomposes) Density (g/mL): 4.5


  • -325 Mesh Powder

Formula: Mn₂O₃
MW: 157.87 g/mol
Melting Pt: 1080 °C (dec.)
Density: 4.5 g/mL
MDL Number: MFCD00016217
CAS Number: 1317-34-6
EINECS: 215-264-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR