3-Amino-5-methyl-1H-pyrazole ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine

126885-100G 126885-25G 126885-5G
BT126885-100GEA 240 USD
BT126885-100G BT126885-25G BT126885-5G
3-Amino-5-methyl-1H-pyrazole ≥97%
3-Amino-5-methyl-1H-pyrazole

CAS: 31230-17-8; MDL No: MFCD00075180 Solid; Molecular Formula: C4H7N3; MW: 97.12 Melting Point: 45-47°; Boiling Point: 213°/14 mmHg; Flash point: 110°C (230°F) Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 213 °C/14 mmHg
Melting Pt: 45…47 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00075180
CAS Number: 31230-17-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR