Methyl arachidate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyl Arachidate
, Methyl eicosanoate
126870-5G
126870-1G
BT126870-5GEA
107.15
USD
BT126870-5G
BT126870-1G
Methyl arachidate ≥98%
Methyl arachidate
- Solid
Formula:
C₂₁H₄₂O₂ MW: 326.56 g/mol Boiling Pt: 215 °C/10 mmHg Melting Pt: 45…48 °C Flash Pt: >110 °C (>230 °F) |
EINECS:
214-304-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...