Methyl-2-aminonicotinate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Methyl 2-aminonicotinate
126815-1G
126815-5G
BT126815-1GEA
117
USD
BT126815-1G
BT126815-5G
Methyl-2-aminonicotinate ≥97%
Methyl-2-aminonicotinate
CAS: 14667-47-1; MDL No: MFCD00128872
Solid; Molecular Formula: C7H8N2O2; MW: 152.15
Melting Point: 82-86°
- Solid
Formula:
C₇H₈N₂O₂ Melting Pt: 82…86 °C |
MDL Number:
MFCD00128872 CAS Number: 14667-47-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...