n-Hexadecane ≥99%
Supplier: BeanTown Chemical
126805-100ML
126805-2.5L
126805-500ML
BT126805-100MLEA
48.97
USD
BT126805-100ML
BT126805-2.5L
BT126805-500ML
n-Hexadecane ≥99%
n-Hexadecane
CAS: 544-76-3; EC No: 208-878-9; MDL No: MFCD00008998; RTECS: ML9200000
Liquid; Linear Formula: CH3(CH2)14CH3; Molecular Formula: C16H34; MW: 226.45
Melting Point: 18°; Boiling Point: 287°; Flash point: 135°C (275°F)
Density (g/mL): 0.773; Refractive Index: 1.434
Hygroscopic
- RTECS: ML9200000
- Refractive index: 1.434
- Hygroscopic
- Liquid
Formula:
H₃C(CH₂)₁₄CH₃ Boiling Pt: 287 °C Melting Pt: 18 °C Density: 0.773 g/mL Flash Pt: 135 °C (275 °F) |
MDL Number:
MFCD00008998 CAS Number: 544-76-3 EINECS: 208-878-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...