Magnesium hydroxide carbonate tetrahydrate

Supplier: BeanTown Chemical

Synonyms: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic

126790-100G 126790-500G
BT126790-100GEA 23 USD
BT126790-100G BT126790-500G
Magnesium hydroxide carbonate tetrahydrate
Magnesium hydroxide carbonate tetrahydrate

CAS: 39409-82-0; EC No: 235-192-7; MDL No: MFCD00149783; RTECS: FG1797000 -325 Mesh Powder; Linear Formula: 4MgCO3·Mg(OH)2·4H2O ; MW: 467.67


  • RTECS: FG1797000
  • -325 Mesh Powder

Formula: 4MgCO₃·Mg(OH)₂·4H₂O
MW: 467.67 g/mol (395.58 anhy)
MDL Number: MFCD00149783
CAS Number: 235-192-7
EINECS: 235-192-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR