2-Chloro-3-formylpyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde

126785-1G 126785-25G 126785-5G
BT126785-1GEA 40 USD
BT126785-1G BT126785-25G BT126785-5G
2-Chloro-3-formylpyridine ≥98%
2-Chloro-3-formylpyridine

CAS: 36404-88-3; MDL No: MFCD01315308 Solid; Molecular Formula: C6H4ClNO; MW: 141.56 Melting Point: 50-54°; Flash point: 110°C (230°F)


  • Solid

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 50…54 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD01315308
CAS Number: 36404-88-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR