o-Cresolphthalein
Supplier: BeanTown Chemical
Synonyms:
3,3-Bis(4-hydroxy-3-methylphenyl)isobenzofuran-1(3H)-one
126720-10G
126720-50G
BT126720-10GEA
30.59
USD
BT126720-10G
BT126720-50G
o-Cresolphthalein
o-Cresolphthalein
CAS: 596-27-0; EC No: 209-881-8; MDL No: MFCD00005912; RTECS: SM8390000
Powder; Molecular Formula: C22H18O4; MW: 346.38
Melting Point: 219-221°
- RTECS: SM8390000
- Powder
Formula:
C₂₂H₁₈O₄ MW: 346.39 g/mol Melting Pt: 219…221 °C |
MDL Number:
MFCD00005912 CAS Number: 596-27-0 EINECS: 209-881-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...