Methyl picolinate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Methyl pyridine-2-carboxylate
126715-100G
126715-25G
BT126715-100GEA
384
USD
BT126715-100G
BT126715-25G
Methyl picolinate ≥99%
Methyl picolinate
CAS: 2459-07-6; EC No: 219-545-2; MDL No: MFCD00038038
Liquid; Molecular Formula: C7H7NO2; MW: 137.14
Boiling Point: 95°/1 mmHg; Flash point: 103°C (217°F)
Density (g/mL): 1.137; Refractive Index: 1.521
- Refractive index: 1.521
- Liquid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 95 °C/1 mmHg Density: 1.137 g/mL Flash Pt: 103 °C (217 °F) |
MDL Number:
MFCD00038038 CAS Number: 2459-07-6 EINECS: 219-545-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...