Yttrium aluminum oxide ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
126710-100G
126710-25G
BT126710-100GEA
709
USD
BT126710-100G
BT126710-25G
Yttrium aluminum oxide ≥99.9% (trace rare earth metals basis)
Yttrium aluminum oxide
CAS: 12005-21-9; EC No: 234-465-8; MDL No: MFCD00213958
Powder; Linear Formula: Y3Al5O12; MW: 593.62
Melting Point: 1760°
Density (g/mL): 4.56; Refractive Index: 1.833
- Refractive index: 1.833
- Powder
Formula:
Y₃Al₅O₁₂ Melting Pt: 1760 °C Density: 4.56 g/mL |
MDL Number:
MFCD00213958 CAS Number: 12005-21-9 EINECS: 234-465-8 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...