5-Amino-2-chloropyridine ≥98%
Supplier: BeanTown Chemical
126705-1G
126705-5G
BT126705-1GEA
59
USD
BT126705-1G
BT126705-5G
5-Amino-2-chloropyridine ≥98%
5-Amino-2-chloropyridine
CAS: 5350-93-6; EC No: 226-322-3; MDL No: MFCD00006243
Crystalline; Molecular Formula: C5H5ClN2; MW: 128.56
Melting Point: 81-83°
- Crystalline
Formula:
C₅H₅ClN₂ Melting Pt: 81…83 °C |
MDL Number:
MFCD00006243 CAS Number: 5350-93-6 EINECS: 226-322-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...