1-(2-Hydroxyethyl)piperazine ≥98%

Supplier: BeanTown Chemical

Synonyms: N-(2-Hydroxyethyl)piperazine, 2-(1-Piperazinyl)ethanol, 2-Piperazin-1-ylethanol

126700-100G 126700-500G
BT126700-100GEA 33 USD
BT126700-100G BT126700-500G
1-(2-Hydroxyethyl)piperazine ≥98%
1-(2-Hydroxyethyl)piperazine

CAS: 103-76-4; EC No: 203-142-3; MDL No: MFCD00005970; RTECS: TL6825000 Liquid; Molecular Formula: C6H14N2O; MW: 130.19 Boiling Point: 246°; Flash point: 135°C (275°F) Density (g/mL): 1.061; Refractive Index: 1.506


  • RTECS: TL6825000
  • Refractive index: 1.506
  • Liquid

Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Boiling Pt: 246 °C
Density: 1.061 g/mL
Flash Pt: 135 °C (275 °F)
MDL Number: MFCD00005970
CAS Number: 103-76-4
EINECS: 203-142-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR