Choline chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

126685-100G 126685-2.5KG 126685-500G
BT126685-100GEA 29.58 USD
BT126685-100G BT126685-2.5KG BT126685-500G
Choline chloride ≥98%
Choline chloride

CAS: 67-48-1; EC No: 200-655-4; MDL No: MFCD00011721; RTECS: KH2975000 Solid; Linear Formula: HOCH2CH2N(CH3)3Cl; Molecular Formula: C5H14ClNO; MW: 139.63 Melting Point: 302-305° (decomposes) Hygroscopic


  • RTECS: KH2975000
  • Hygroscopic
  • Solid

Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
Melting Pt: 302…305 °C (dec.)
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR