1,3-Bis(diphenylphosphino)propane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane
126680-100G
126680-25G
126680-5G
BT126680-100GEA
694
USD
BT126680-100G
BT126680-25G
BT126680-5G
1,3-Bis(diphenylphosphino)propane ≥97%
1,3-Bis(diphenylphosphino)propane
CAS: 6737-42-4; EC No: 229-791-2; MDL No: MFCD00003050
Crystalline; Molecular Formula: C27H26P2; MW: 412.44
Melting Point: 63-65°
Air Sensitive
- Air Sensitive
- Crystalline
Formula:
C₂₇H₂₆P₂ MW: 412.45 g/mol Melting Pt: 63…65 °C |
MDL Number:
MFCD00003050 CAS Number: 6737-42-4 EINECS: 229-791-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...