Vincamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Vincamine base, Methyl (41S,12S,13aS)-13a-ethyl-12-hydroxy-2,3,41,5,6,12,13,13a-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate
126670-1G
126670-5G
BT126670-1GEA
112
USD
BT126670-1G
BT126670-5G
Vincamine ≥98%
Vincamine
CAS: 1617-90-9; EC No: 216-576-3; MDL No: MFCD00078054; RTECS: YY8575000
Crystalline/Powder; Molecular Formula: C21H26N2O3; MW: 354.44
Melting Point: 232° (decomposes)
Optical Rotation: [α]23/D +42.8°, c = 1 in pyridine
- RTECS: YY8575000
- Optical rotation: [α]23/D +42.8°, c = 1 in pyridine
- Crystalline/Powder
Formula:
C₂₁H₂₆N₂O₃ MW: 354.45 g/mol Melting Pt: 232 °C (dec.) |
MDL Number:
MFCD00078054 CAS Number: 1617-90-9 EINECS: 216-576-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...