Aluminium oxide, α-phase ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
126665-100G
126665-25G
BT126665-100GEA
263
USD
BT126665-100G
BT126665-25G
Aluminium oxide, α-phase ≥99.99% (trace metals basis)
Aluminium oxide, α-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
2-12mm Pieces ; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- 2-12mm Pieces
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...