Thiamine (Vitamin B1)
Supplier: BeanTown Chemical
Synonyms:
Thiamine monochloride, Aneurine, Vitamin B1, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride
126650-1G
126650-5G
BT126650-1GEA
65.4
USD
BT126650-1G
BT126650-5G
Thiamine (Vitamin B1)
Thiamine (Vitamin B1)
CAS: 59-43-8; EC No: 200-425-3; MDL No: MFCD00012780
Solid; Molecular Formula: C12H17ClN4OS; MW: 300.81
- Solid
Formula:
C₁₂H₁₇ClN₄OS |
MDL Number:
MFCD00044586 CAS Number: 59-43-8 EINECS: 200-425-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...