N-Boc-L-alaninol ≥99%
Supplier: BeanTown Chemical
Synonyms:
Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol
126550-100G
126550-25G
126550-5G
BT126550-100GEA
1748
USD
BT126550-100G
BT126550-25G
BT126550-5G
N-Boc-L-alaninol ≥99%
N-Boc-L-alaninol
CAS: 79069-13-9; MDL No: MFCD00043121
Crystalline; Molecular Formula: C8H17NO3 ; MW: 175.23
Melting Point: 59-62°
Optical Rotation: [α]20/D -11°, c = 1 in chloroform
- Optical rotation: [α]20/D -11°, c = 1 in chloroform
- Crystalline
Formula:
C₈H₁₇NO₃ Melting Pt: 59…62 °C |
MDL Number:
MFCD00043121 CAS Number: 79069-13-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...