(Triphenylphosphine)gold(I) chloride ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

144720-1G 144720-500MG 144720-5G
BT144720-1GEA 208.44 USD
BT144720-1G BT144720-500MG BT144720-5G
(Triphenylphosphine)gold(I) chloride ≥99.99%
(Triphenylphosphine)gold(I) chloride

CAS: 14243-64-2; EC No: 238-117-6; MDL No: MFCD00009588 Powder; Linear Formula: AuCl[P(C6H5)3]; Molecular Formula: C18H15AuClP; MW: 494.71


  • Powder

Formula: C₁₈H₁₅AuClP
MW: 494.71
MDL Number: MFCD00009588
CAS Number: 14243-64-2
EINECS: 238-117-6

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR