trans,trans-Dibenzylideneacetone ≥98%
Supplier: BeanTown Chemical
Synonyms:
(1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one
144670-100G
144670-25G
144670-5G
BT144670-100GEA
232
USD
BT144670-100G
BT144670-25G
BT144670-5G
trans,trans-Dibenzylideneacetone ≥98%
trans,trans-Dibenzylideneacetone
CAS: 35225-79-7; EC No: 208-697-5; MDL No: MFCD00004790
Solid; Linear Formula: C6H5CH=CHCOCH=CHC6H5; Molecular Formula: C17H14O; MW: 234.30
Melting Point: 112-114°
- Solid
Formula:
C₁₇H₁₄O MW: 234.30 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD00004790 CAS Number: 35225-79-7 EINECS: 208-697-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...