2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-4,6-dimethoxy-s-triazine
144640-100G
144640-25G
144640-5G
BT144640-100GEA
338
USD
BT144640-100G
BT144640-25G
BT144640-5G
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine
CAS: 3140-73-6; EC No: 221-541-0; MDL No: MFCD00075607
Crystalline/Powder; Molecular Formula: C5H6ClN3O2; MW: 175.57
Melting Point: 71-74°
- Crystalline/Powder
Formula:
C₅H₆ClN₃O₂ Melting Pt: 71…74 °C |
MDL Number:
MFCD00075607 CAS Number: 3140-73-6 EINECS: 221-541-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...