Ethyl-3,5-dimethylisoxazole-4-carboxylate ≥97%
Supplier: BeanTown Chemical
144615-1G
144615-5G
BT144615-1GEA
26
USD
BT144615-1G
BT144615-5G
Ethyl-3,5-dimethylisoxazole-4-carboxylate ≥97%
CAS: 17147-42-1; MDL No: MFCD00052557
Liquid; Molecular Formula: C8H11NO3; MW: 169.18
Boiling Point: 87-89°/2 mmHg
- Liquid
Formula:
C₈H₁₁NO₃ Boiling Pt: 87…89 °C/2 mmHg |
MDL Number:
MFCD00052557 CAS Number: 17147-42-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...