Ammonium benzenesulfonate

Supplier: BeanTown Chemical

144600-1G 144600-5G
BT144600-1GEA 49 USD
BT144600-1G BT144600-5G
Ammonium benzenesulfonate
Ammonium benzenesulfonate

CAS: 19402-64-3; EC No: 243-035-9 Solid; Molecular Formula: C6H9NO3S; MW: 175.21


  • Solid

Formula: C₆H₉NO₃S
MW: 175.2100 g/mol
MDL Number: MFCD00191294
CAS Number: 19402-64-3
EINECS: 243-035-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR