N-(Chloroacetyl)-3-(trifluoromethyl)aniline
Supplier: BeanTown Chemical
144580-1G
144580-5G
BT144580-1GEA
105
USD
BT144580-1G
BT144580-5G
N-(Chloroacetyl)-3-(trifluoromethyl)aniline
N-(Chloroacetyl)-3-(trifluoromethyl)aniline
CAS: 351-38-2; MDL No: MFCD00084932
Solid; Molecular Formula: C9H7ClF3NO; MW: 237.61
Melting Point: 62.5-64°
- Solid
Formula:
C₉H₇ClF₃NO Melting Pt: 62.5…64 °C |
CAS Number:
351-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...