4,4'-(Hexafluoroisopropylidene)diphenol ≥98%
Supplier: BeanTown Chemical
144555-100G
144555-25G
144555-5G
BT144555-100GEA
389
USD
BT144555-100G
BT144555-25G
BT144555-5G
4,4'-(Hexafluoroisopropylidene)diphenol ≥98%
4,4'-(Hexafluoroisopropylidene)diphenol
CAS: 1478-61-1; EC No: 216-036-7; MDL No: MFCD00000439; RTECS: SN2780000
Powder; Molecular Formula: C15H10F6O2; MW: 336.23
Melting Point: 160-163°
- RTECS: SN2780000
- Powder
Formula:
C₁₅H₁₀F₆O₂ Melting Pt: 160…163 °C |
MDL Number:
MFCD00000439 CAS Number: 1478-61-1 EINECS: 216-036-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...