2-Bromo-5-nitropyridine ≥98%

Supplier: BeanTown Chemical

135440-1G 135440-5G
BT135440-1GEA 27 USD
BT135440-1G BT135440-5G
2-Bromo-5-nitropyridine ≥98%
2-Bromo-5-nitropyridine

CAS: 4487-59-6; EC No: 224-777-2; MDL No: MFCD00006222 Crystalline; Molecular Formula: C5H3BrN2O2 ; MW: 202.99 Melting Point: 139-141°; Boiling Point: 145-147°/10 mmHg


  • Crystalline

Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Boiling Pt: 145…147 °C/10 mmHg
Melting Pt: 139…141 °C
MDL Number: MFCD00006222
CAS Number: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR