2-Pyrrolecarbaldehyde ≥99%
Supplier: BeanTown Chemical
Synonyms:
1H-Pyrrole-2-carboxaldehyde, Pyrrole-2-carboxaldehyde, 1H-Pyrrole-2-carbaldehyde, Pyrrole-2-carbaldehyde
135420-100G
135420-25G
135420-5G
BT135420-100GEA
389
USD
BT135420-100G
BT135420-25G
BT135420-5G
2-Pyrrolecarbaldehyde ≥99%
2-Pyrrolecarbaldehyde
CAS: 1003-29-8; EC No: 213-705-5; MDL No: MFCD00005217
Crystalline; Molecular Formula: C5H5NO; MW: 95.10
Melting Point: 43-46°; Boiling Point: 217-219°; Flash point: 106°C (222°F)
Air Sensitive
- Air Sensitive
- Crystalline
Formula:
C₅H₅NO MW: 95.1 g/mol Boiling Pt: 217…219 °C Melting Pt: 43…46 °C Flash Pt: 106 °C (222 °F) |
MDL Number:
MFCD00005217 CAS Number: 1003-29-8 EINECS: 213-705-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...