11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one ≥98%
Supplier: BeanTown Chemical
135380-1G
135380-5G
BT135380-1GEA
37
USD
BT135380-1G
BT135380-5G
11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one ≥98%
11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one
CAS: 28797-48-0; EC No: 249-227-9
Solid; Molecular Formula: C14H12ClN3O2; MW: 287.70
Boiling Point: 481°
- Solid
Formula:
C₁₄H₁₀ClN₃O₂ Boiling Pt: 481 °C |
MDL Number:
MFCD21648226 CAS Number: 28797-48-0 EINECS: 249-227-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...